Assay Details
Gene Symbol: dhrs13b.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083830.1 | NP_001077299.1 | 3 - 4 | 596 | 75 | |
GenBank mRNA | BC171514.1 | - | 3 - 4 | 577 | 75 | |
BC171516.1 | - | 3 - 4 | 577 | 75 | ||
CU458961.1 | - | 3 - 4 | 596 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
