Assay Details
Gene Symbol: igf2bp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089374.1 | NP_001082843.1 | 5 - 6 | 837 | 64 | |
XM_005159948.4 | XP_005160005.1 | 6 - 7 | 949 | 64 | ||
XM_005159949.4 | XP_005160006.1 | 6 - 7 | 1113 | 64 | ||
XM_017354915.2 | XP_017210404.1 | 6 - 7 | 1182 | 64 | ||
XM_021474969.1 | XP_021330644.1 | 6 - 7 | 1111 | 64 | ||
GenBank mRNA | BC134152.1 | - | 3 - 4 | 312 | 64 | |
EU296399.1 | - | 7 - 8 | 1068 | 64 | ||
EU296400.1 | - | 6 - 7 | 883 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
