Assay Details
| Gene Symbol: mfsd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089520.1 | NP_001082989.1 | 3 - 4 | 801 | 65 | |
| GenBank mRNA | BC134911.1 | - | 3 - 4 | 801 | 65 | |
More Information
Set Membership: |
|

mfsd4b
Genomic Map