Assay Details
| Gene Symbol: gtf3ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089544.2 | NP_001083013.2 | 1 - 2 | 247 | 81 | |
| GenBank mRNA | BC133956.1 | - | 1 - 2 | 175 | 81 | |
| EH546778.1 | - | 1 - 2 | 216 | 81 | ||
More Information
Set Membership: |
|

gtf3ab
Genomic Map