Assay Details
| Gene Symbol: gad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089570.2 | NP_001083039.2 | 11 - 12 | 1168 | 101 | |
| GenBank mRNA | BC135107.1 | - | 10 - 11 | 1016 | 101 | |
More Information
Set Membership: |
|

gad3
Genomic Map