Assay Details
Gene Symbol: ppp4r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110371.1 | NP_001103841.1 | 2 - 3 | 208 | 134 | |
GenBank mRNA | BC154497.1 | - | 2 - 3 | 208 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
