Assay Details
Gene Symbol: ppp4r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110371.1 | NP_001103841.1 | 5 - 6 | 510 | 140 | |
GenBank mRNA | BC096878.1 | - | 5 - 6 | 484 | 140 | |
BC129134.1 | - | 5 - 6 | 494 | 140 | ||
BC154497.1 | - | 5 - 6 | 510 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
