Assay Details
| Gene Symbol: shda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110835.1 | NP_001104305.1 | 5 - 6 | 839 | 65 | |
| GenBank mRNA | BC097175.1 | - | 5 - 6 | 1035 | 65 | |
More Information
Set Membership: |
|

shda
Genomic Map