Assay Details
| Gene Symbol: il13ra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110837.1 | NP_001104307.1 | 7 - 8 | 1102 | 60 | |
| GenBank mRNA | BC162934.1 | - | 7 - 8 | 1105 | 60 | |
More Information
Set Membership: |
|

il13ra1
Genomic Map