Assay Details
Gene Symbol: ghrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111081.2 | NP_001104551.1 | 2 - 3 | 468 | 138 | |
GenBank mRNA | EU649775.1 | - | 2 - 3 | 471 | 141 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001111081.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
