Assay Details
Gene Symbol: ghrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111081.2 | NP_001104551.1 | 7 - 8 | 1179 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
