Assay Details
| Gene Symbol: abcg4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111212.1 | NP_001104682.1 | 11 - 12 | 1376 | 71 | |
| GenBank mRNA | BC153997.1 | - | 11 - 12 | 1376 | 71 | |
More Information
Set Membership: |
|

abcg4b
Genomic Map