Assay Details
| Gene Symbol: socs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111225.1 | NP_001104695.1 | 1 | 1047 | 103 | |
| XM_021467245.1 | XP_021322920.1 | 2 | 1390 | 103 | ||
| GenBank mRNA | EF371367.1 | - | 1 | 1047 | 103 | |
| FR750002.1 | - | 1 | 1047 | 103 | ||
More Information
Set Membership: |
|

socs4
Genomic Map