Assay Details
| Gene Symbol: hbegfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111226.1 | NP_001104696.1 | 2 - 3 | 377 | 77 | |
| GenBank mRNA | BC155826.1 | - | 2 - 3 | 377 | 77 | |
More Information
Set Membership: |
|

hbegfa
Genomic Map