Assay Details
| Gene Symbol: kcnk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111231.1 | NP_001104701.1 | 1 - 2 | 451 | 85 | |
| XM_005161888.4 | XP_005161945.1 | 1 - 2 | 631 | 85 | ||
| XM_005161889.4 | XP_005161946.1 | 1 - 2 | 652 | 85 | ||
| GenBank mRNA | BC153969.1 | - | 1 - 2 | 451 | 85 | |
More Information
Set Membership: |
|

kcnk15
Genomic Map