Assay Details
| Gene Symbol: ctps1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111247.1 | NP_001104717.1 | 16 - 17 | 1751 | 87 | |
| GenBank mRNA | BC155167.1 | - | 16 - 17 | 1751 | 87 | |
| BC171607.1 | - | 16 - 17 | 1726 | 87 | ||
More Information
Set Membership: |
|

ctps1b
Genomic Map