Assay Details
Gene Symbol: prlrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113500.1 | NP_001106972.1 | 2 - 3 | 212 | 71 | |
XM_005165610.3 | XP_005165667.1 | 3 - 4 | 418 | 71 | ||
GenBank mRNA | EU517720.1 | - | 3 - 4 | 390 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
