Assay Details
Gene Symbol: prlrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113500.1 | NP_001106972.1 | 5 - 6 | 702 | 77 | |
XM_005165610.3 | XP_005165667.1 | 6 - 7 | 908 | 77 | ||
GenBank mRNA | EU517720.1 | - | 6 - 7 | 880 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
