Assay Details
| Gene Symbol: ghrhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113503.1 | NP_001106975.1 | 2 - 3 | 163 | 83 | |
| GenBank mRNA | EU056169.1 | - | 2 - 3 | 163 | 83 | |
More Information
Set Membership: |
|

ghrhr2
Genomic Map