Assay Details
| Gene Symbol: il23r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113506.1 | NP_001106978.1 | 5 - 6 | 795 | 75 | |
| XM_021476745.1 | XP_021332420.1 | 8 - 9 | 1677 | 75 | ||
More Information
Set Membership: |
|

il23r
Genomic Map