Assay Details
| Gene Symbol: cadm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113552.1 | NP_001107024.1 | 9 - 10 | 1156 | 61 | |
| GenBank mRNA | EU182352.1 | - | 9 - 10 | 1156 | 61 | |
More Information
Set Membership: |
|

cadm1b
Genomic Map