Assay Details
| Gene Symbol: paox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113603.1 | NP_001107075.1 | 1 - 2 | 64 | 69 | |
| XM_005166871.4 | XP_005166928.1 | 3 - 4 | 433 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001113603.1, XM_005166871.4 transcripts.
Set Membership: |
|

paox1
Genomic Map