Assay Details
| Gene Symbol: paox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113603.1 | NP_001107075.1 | 7 - 8 | 1396 | 58 | |
| XM_005166871.4 | XP_005166928.1 | 9 - 10 | 1765 | 58 | ||
More Information
Set Membership: |
|

paox1
Genomic Map