Assay Details
Gene Symbol: lifrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113732.1 | NP_001107204.1 | 17 - 18 | 2378 | 66 | |
XM_009295324.3 | XP_009293599.1 | 17 - 18 | 2433 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
