Assay Details
Gene Symbol: lifrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113732.1 | NP_001107204.1 | 5 - 6 | 844 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
