Assay Details
Gene Symbol: lifrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113732.1 | NP_001107204.1 | 7 - 8 | 1091 | 114 | |
XM_009295324.3 | XP_009293599.1 | 7 - 8 | 1134 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
