Assay Details
| Gene Symbol: ca4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114407.2 | NP_001107879.2 | 3 - 4 | 304 | 94 | |
| GenBank mRNA | BC155085.1 | - | 3 - 4 | 304 | 94 | |
More Information
Set Membership: |
|

ca4a
Genomic Map