Assay Details
| Gene Symbol: slc26a6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114417.1 | NP_001107889.1 | 12 - 13 | 1576 | 71 | |
| GenBank mRNA | BC155340.1 | - | 12 - 13 | 1576 | 71 | |
| FJ170818.1 | - | 12 - 13 | 1575 | 71 | ||
More Information
Set Membership: |
|

slc26a6l
Genomic Map