Assay Details
| Gene Symbol: ramp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114432.1 | NP_001107904.1 | 5 - 6 | 406 | 69 | |
| XM_005164138.4 | XP_005164195.1 | 3 - 4 | 462 | 69 | ||
| XM_017354532.2 | XP_017210021.1 | 5 - 6 | 453 | 69 | ||
| GenBank mRNA | BC090460.1 | - | 3 - 4 | 411 | 69 | |
| BC154812.1 | - | 5 - 6 | 406 | 69 | ||
More Information
Set Membership: |
|

ramp2
Genomic Map