Assay Details
| Gene Symbol: slc4a2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114440.1 | NP_001107912.1 | 17 - 18 | 2947 | 102 | |
| GenBank mRNA | DQ286970.1 | - | 17 - 18 | 2947 | 102 | |
| DQ287972.1 | - | 17 - 18 | 2947 | 102 | ||
More Information
Set Membership: |
|

slc4a2b
Genomic Map