Assay Details
| Gene Symbol: msl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114468.1 | NP_001107940.1 | 1 - 2 | 54 | 104 | |
| XM_021468802.1 | XP_021324477.1 | 1 - 2 | 104 | 104 | ||
| GenBank mRNA | BC155827.1 | - | 1 - 2 | 54 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC155827.1, NM_001114468.1, XM_021468802.1 transcripts.
Set Membership: |
|

msl1a
Genomic Map