Assay Details
| Gene Symbol: ndr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130966.1 | NP_571041.1 | 2 - 3 | 1075 | 79 | |
| GenBank mRNA | AF002218.1 | - | 2 - 3 | 1203 | 79 | |
| AF003699.1 | - | 1 - 2 | 401 | 79 | ||
| AF056327.1 | - | 2 - 3 | 1075 | 79 | ||
| BC097089.1 | - | 2 - 3 | 1117 | 79 | ||
More Information
Set Membership: |
|

ndr1
Genomic Map