Assay Details
| Gene Symbol: snai1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130989.3 | NP_571064.2 | 2 - 3 | 623 | 145 | |
| XM_021469851.1 | XP_021325526.1 | 2 - 3 | 729 | 145 | ||
| GenBank mRNA | BC162632.1 | - | 2 - 3 | 623 | 145 | |
| U24225.1 | - | 2 - 3 | 670 | 145 | ||
More Information
Set Membership: |
|

snai1b
Genomic Map