Assay Details
Gene Symbol: ahr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131028.2 | NP_571103.2 | 4 - 5 | 766 | 99 | |
XM_021466612.1 | XP_021322287.1 | 4 - 5 | 783 | 99 | ||
GenBank mRNA | AF258854.1 | - | 4 - 5 | 766 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
