Assay Details
Gene Symbol: ahr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131028.2 | NP_571103.2 | 9 - 10 | 1475 | 71 | |
XM_021466612.1 | XP_021322287.1 | 9 - 10 | 1492 | 71 | ||
GenBank mRNA | AF258854.1 | - | 9 - 10 | 1475 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
