Assay Details
| Gene Symbol: tcf7l1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131296.2 | NP_571371.2 | 5 - 6 | 1251 | 137 | |
| GenBank mRNA | AY221031.1 | - | 5 - 6 | 1252 | 137 | |
More Information
Set Membership: |
|

tcf7l1b
Genomic Map