Assay Details
| Gene Symbol: tcf7l1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131296.2 | NP_571371.2 | 6 - 7 | 1358 | 105 | |
| GenBank mRNA | AY221031.1 | - | 6 - 7 | 1359 | 105 | |
More Information
Set Membership: |
|

tcf7l1b
Genomic Map