Assay Details
| Gene Symbol: tcf7l1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131296.2 | NP_571371.2 | 9 - 10 | 1746 | 59 | |
| GenBank mRNA | AF136456.1 | - | 2 - 3 | 301 | 59 | |
| AY221031.1 | - | 9 - 10 | 1747 | 59 | ||
More Information
Set Membership: |
|

tcf7l1b
Genomic Map