Assay Details
| Gene Symbol: kdrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131472.1 | NP_571547.1 | 10 - 11 | 1404 | 90 | |
| XM_009291142.3 | XP_009289417.1 | 10 - 11 | 1798 | 90 | ||
| GenBank mRNA | AF180354.1 | - | 7 - 8 | 1017 | 90 | |
| AF487829.1 | - | 10 - 11 | 1754 | 90 | ||
| AY056466.1 | - | 10 - 11 | 1404 | 90 | ||
| BC129158.1 | - | 10 - 11 | 1769 | 90 | ||
| CU458916.1 | - | 10 - 11 | 1769 | 90 | ||
More Information
Set Membership: |
|

kdrl
Genomic Map