Assay Details
| Gene Symbol: kdrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131472.1 | NP_571547.1 | 11 - 12 | 1513 | 84 | |
| XM_009291142.3 | XP_009289417.1 | 11 - 12 | 1907 | 84 | ||
| GenBank mRNA | AF180354.1 | - | 8 - 9 | 1129 | 84 | |
| AF487829.1 | - | 11 - 12 | 1866 | 84 | ||
| AY056466.1 | - | 11 - 12 | 1513 | 84 | ||
| BC129158.1 | - | 11 - 12 | 1881 | 84 | ||
| CU458916.1 | - | 11 - 12 | 1881 | 84 | ||
More Information
Set Membership: |
|

kdrl
Genomic Map