Assay Details
| Gene Symbol: kdrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131472.1 | NP_571547.1 | 12 - 13 | 1614 | 70 | |
| XM_009291142.3 | XP_009289417.1 | 12 - 13 | 2008 | 70 | ||
| GenBank mRNA | AF180354.1 | - | 9 - 10 | 1230 | 70 | |
| AF487829.1 | - | 12 - 13 | 1967 | 70 | ||
| AY056466.1 | - | 12 - 13 | 1614 | 70 | ||
| BC129158.1 | - | 12 - 13 | 1982 | 70 | ||
| CU458916.1 | - | 12 - 13 | 1982 | 70 | ||
More Information
Set Membership: |
|

kdrl
Genomic Map