Assay Details
| Gene Symbol: kdrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131472.1 | NP_571547.1 | 17 - 18 | 2430 | 85 | |
| XM_009291142.3 | XP_009289417.1 | 17 - 18 | 2824 | 85 | ||
| GenBank mRNA | AF180354.1 | - | 14 - 15 | 2046 | 85 | |
| AF487829.1 | - | 17 - 18 | 2783 | 85 | ||
| AY056466.1 | - | 17 - 18 | 2430 | 85 | ||
| BC129158.1 | - | 17 - 18 | 2798 | 85 | ||
| CU458916.1 | - | 17 - 18 | 2798 | 85 | ||
More Information
Set Membership: |
|

kdrl
Genomic Map