Assay Details
| Gene Symbol: kdrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131472.1 | NP_571547.1 | 25 - 26 | 3331 | 94 | |
| XM_009291142.3 | XP_009289417.1 | 25 - 26 | 3725 | 94 | ||
| GenBank mRNA | AF180354.1 | - | 22 - 23 | 2947 | 94 | |
| AF487829.1 | - | 25 - 26 | 3684 | 94 | ||
| AY056466.1 | - | 25 - 26 | 3331 | 94 | ||
| BC129158.1 | - | 25 - 26 | 3699 | 94 | ||
| CU458916.1 | - | 25 - 26 | 3699 | 94 | ||
| U75995.1 | - | 4 - 5 | 403 | 94 | ||
| U82383.1 | - | 5 - 6 | 457 | 94 | ||
| U89515.1 | - | 7 - 8 | 760 | 94 | ||
More Information
Set Membership: |
|

kdrl
Genomic Map