Assay Details
| Gene Symbol: kdrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131472.1 | NP_571547.1 | 7 - 8 | 959 | 132 | |
| XM_009291142.3 | XP_009289417.1 | 7 - 8 | 1353 | 132 | ||
| GenBank mRNA | AF180354.1 | - | 4 - 5 | 572 | 132 | |
| AF487829.1 | - | 7 - 8 | 1309 | 132 | ||
| AY056466.1 | - | 7 - 8 | 959 | 132 | ||
| BC129158.1 | - | 7 - 8 | 1324 | 132 | ||
| CU458916.1 | - | 7 - 8 | 1324 | 132 | ||
More Information
Set Membership: |
|

kdrl
Genomic Map