Assay Details
| Gene Symbol: smx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131496.2 | NP_571571.1 | 2 - 3 | 142 | 144 | |
| GenBank mRNA | BC085441.1 | - | 2 - 3 | 104 | 144 | |
| BC152150.1 | - | 2 - 3 | 129 | 144 | ||
| BC164435.1 | - | 1 - 2 | 100 | 144 | ||
| EH449634.1 | - | 2 - 3 | 144 | 144 | ||
More Information
Set Membership: |
|

smx5
Genomic Map