Assay Details
| Gene Symbol: cpeb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131725.1 | NP_571800.1 | 1 - 2 | 119 | 91 | |
| GenBank mRNA | AB011681.1 | - | 1 - 2 | 119 | 91 | |
| BC163653.1 | - | 2 - 3 | 216 | 91 | ||
| BC163656.1 | - | 2 - 3 | 216 | 91 | ||
More Information
Set Membership: |
|

cpeb1a
Genomic Map