Assay Details
| Gene Symbol: cpeb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131725.1 | NP_571800.1 | 9 - 10 | 1379 | 130 | |
| XM_009297761.3 | XP_009296036.1 | 9 - 10 | 1506 | 130 | ||
| GenBank mRNA | AB011681.1 | - | 9 - 10 | 1379 | 130 | |
| BC163653.1 | - | 10 - 11 | 1476 | 130 | ||
| BC163656.1 | - | 10 - 11 | 1476 | 130 | ||
More Information
Set Membership: |
|

cpeb1a
Genomic Map