Assay Details
Gene Symbol: efnb2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131808.1 | NP_571883.1 | 3 - 4 | 498 | 87 | |
GenBank mRNA | AF375226.1 | - | 3 - 4 | 498 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
