Assay Details
Gene Symbol: klf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131859.3 | NP_571934.3 | 2 - 3 | 238 | 74 | |
XM_017356404.2 | XP_017211893.1 | 2 - 3 | 347 | 74 | ||
GenBank mRNA | AF392993.1 | - | 2 - 3 | 245 | 74 | |
BC066635.1 | - | 2 - 3 | 209 | 74 | ||
CK025353.1 | - | 2 - 3 | 185 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
