Assay Details
Gene Symbol: cygb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152952.2 | NP_694484.1 | 2 - 3 | 493 | 90 | |
GenBank mRNA | AJ320232.1 | - | 2 - 3 | 496 | 90 | |
BC094999.1 | - | 2 - 3 | 493 | 90 | ||
BC165894.1 | - | 2 - 3 | 402 | 90 | ||
EH436870.1 | - | 2 - 3 | 522 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
